siRNA Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198752.3 | NP_942047.2 | 11, 12 | 1257 | ||
| GenBank mRNA | AJ250329.1 | - | 11, 12 | 1257 | ||
| AJ250330.1 | - | 10, 11 | 1298 | |||
| FQ211833.1 | - | 9, 10 | 1221 | |||

Kifc2
Genomic Map