siRNA Details
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106788.1 | NP_001100258.1 | 4, 5 | 374 | ||
XM_006242182.2 | XP_006242244.1 | 4, 5 | 936 | |||
XM_006242183.2 | XP_006242245.1 | 4, 5 | 789 |
