siRNA Details
| Gene Symbol: Troap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271243.1 | NP_001258172.1 | 13 | 1939 | ||
| XM_006257348.2 | XP_006257410.1 | 12 | 1832 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Troap| Gene Symbol: Troap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271243.1 | NP_001258172.1 | 13 | 1939 | ||
| XM_006257348.2 | XP_006257410.1 | 12 | 1832 | |||