siRNA Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191614.1 | NP_001178543.1 | 6 | 768 | ||
| XM_006257350.1 | XP_006257412.1 | 7 | 1190 | |||
| XM_006257351.2 | XP_006257413.1 | 7 | 946 | |||
| XM_008765701.1 | XP_008763923.1 | 7 | 2836 | |||
| XM_008765702.1 | XP_008763924.1 | 7 | 1247 | |||

Spats2
Genomic Map