siRNA Details
| Gene Symbol: Mcrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013106.1 | NP_001013124.1 | 4 | 455 | ||
| GenBank mRNA | BC091366.1 | - | 4 | 455 | 7319183 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcrs1| Gene Symbol: Mcrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013106.1 | NP_001013124.1 | 4 | 455 | ||
| GenBank mRNA | BC091366.1 | - | 4 | 455 | 7319183 | |