siRNA Details
| Gene Symbol: Cbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106797.3 | NP_001100267.1 | 3 | 270 | ||
| XM_006242391.2 | XP_006242453.1 | 3 | 334 | |||
| XM_006242393.2 | XP_006242455.1 | 2 | 229 | |||
| XM_008765707.1 | XP_008763929.1 | 2 | 243 | |||
| GenBank mRNA | BC166908.1 | - | 3 | 271 | 6889335 | |
| BC168739.1 | - | 3 | 271 | 6918617 | ||

Cbx5
Genomic Map