siRNA Details
| Gene Symbol: Nnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106819.1 | NP_001100289.1 | 3 | 588 | ||
| XM_008766195.1 | XP_008764417.1 | 3 | 813 | |||
| GenBank mRNA | FQ218983.1 | - | 3 | 552 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nnmt| Gene Symbol: Nnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106819.1 | NP_001100289.1 | 3 | 588 | ||
| XM_008766195.1 | XP_008764417.1 | 3 | 813 | |||
| GenBank mRNA | FQ218983.1 | - | 3 | 552 | ||