siRNA Details
| Gene Symbol: Lman1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012465.1 | NP_001012483.1 | 6 | 640 | ||
| GenBank mRNA | AB188302.1 | - | 6 | 640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lman1l| Gene Symbol: Lman1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012465.1 | NP_001012483.1 | 6 | 640 | ||
| GenBank mRNA | AB188302.1 | - | 6 | 640 | ||