siRNA Details
| Gene Symbol: Fam214a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106838.1 | NP_001100308.1 | 7 | 2354 | ||
| XM_008766246.1 | XP_008764468.1 | 6 | 4361 | |||
| GenBank mRNA | FQ212313.1 | - | 6 | 2192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam214a| Gene Symbol: Fam214a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106838.1 | NP_001100308.1 | 7 | 2354 | ||
| XM_008766246.1 | XP_008764468.1 | 6 | 4361 | |||
| GenBank mRNA | FQ212313.1 | - | 6 | 2192 | ||