siRNA Details
| Gene Symbol: Lysmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106839.1 | NP_001100309.1 | 2 | 671 | ||
| XM_006243405.2 | XP_006243467.1 | 2 | 1564 | |||
| XM_008766248.1 | XP_008764470.1 | 3 | 531 | |||
| GenBank mRNA | BC167017.1 | - | 2 | 563 | 7302522 | |

Lysmd2
Genomic Map