siRNA Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004250.2 | NP_001004250.1 | 7, 8 | 812 | ||
| GenBank mRNA | BC078923.1 | - | 7, 8 | 814 | 7106265 | |
| FQ213124.1 | - | 7, 8 | 820 | |||
| FQ215592.1 | - | 3, 4 | 296 | |||
| FQ224157.1 | - | 7, 8 | 820 | |||

Uqcrc1
Genomic Map