siRNA Details
| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198754.2 | NP_942049.1 | 2 | 543 | ||
| XM_006243964.2 | XP_006244026.1 | 2 | 249 | |||
| GenBank mRNA | AF538323.1 | - | 2 | 346 | ||
| BC088094.1 | - | 2 | 543 | 7325894 | ||

Cmtm8
Genomic Map