siRNA Details
| Gene Symbol: Slc25a41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106874.1 | NP_001100344.1 | 7 | 908 | ||
| XM_006244305.1 | XP_006244367.1 | 7 | 957 | |||
| XM_008766754.1 | XP_008764976.1 | 7 | 1018 | |||
| GenBank mRNA | FM165287.1 | - | 7 | 845 | ||

Slc25a41
Genomic Map