siRNA Details
| Gene Symbol: Cnnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106901.2 | NP_001100371.2 | 4 | 1710 | ||
| XM_006244756.2 | XP_006244818.1 | 4 | 1717 | |||
| XM_006244757.2 | XP_006244819.1 | 4 | 1720 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnnm3| Gene Symbol: Cnnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106901.2 | NP_001100371.2 | 4 | 1710 | ||
| XM_006244756.2 | XP_006244818.1 | 4 | 1717 | |||
| XM_006244757.2 | XP_006244819.1 | 4 | 1720 | |||