siRNA Details
| Gene Symbol: Arpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106919.1 | NP_001100389.1 | 9 | 869 | ||
| XM_006245165.1 | XP_006245227.1 | 8 | 1007 | |||
| XM_006245166.1 | XP_006245228.1 | 9 | 824 | |||
| GenBank mRNA | FQ214133.1 | - | 9 | 830 | ||
| FQ226423.1 | - | 3 | 269 | |||
| FQ231947.1 | - | 6 | 622 | |||

Arpc2
Genomic Map