siRNA Details
| Gene Symbol: Pof1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106934.1 | NP_001100404.1 | 7 | 909 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pof1b| Gene Symbol: Pof1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106934.1 | NP_001100404.1 | 7 | 909 | ||