siRNA Details
Gene Symbol: C1galt1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030033.1 | NP_001025204.1 | 2 | 397 | ||
XM_006257487.1 | XP_006257549.1 | 2 | 446 | |||
GenBank mRNA | BC099818.1 | - | 2 | 397 | 7930810 |
