siRNA Details
| Gene Symbol: Glod5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106957.1 | NP_001100427.1 | 3 | 328 | ||
| XM_006256706.2 | XP_006256768.1 | 4 | 381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glod5| Gene Symbol: Glod5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106957.1 | NP_001100427.1 | 3 | 328 | ||
| XM_006256706.2 | XP_006256768.1 | 4 | 381 | |||