siRNA Details
| Gene Symbol: Sat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007667.2 | NP_001007668.1 | 6 | 677 | ||
| XM_008773228.1 | XP_008771450.1 | 6 | 1677 | |||
| XM_008773229.1 | XP_008771451.1 | 6 | 1073 | |||
| XM_008773230.1 | XP_008771452.1 | 7 | 712 | |||
| GenBank mRNA | AY325208.1 | - | 542 | |||
| BC060588.1 | - | 6 | 679 | 6921572 | ||
| FQ222039.1 | - | 6 | 559 | |||
| FQ229984.1 | - | 6 | 561 | |||

Sat1
Genomic Map