siRNA Details
| Gene Symbol: Olr1765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000489.1 | NP_001000489.1 | 1 | 553 | ||
| Gene Symbol: LOC103689939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773554.1 | XP_008771776.1 | 322 | |||
More Information
Set Membership: |
|

Olr1765
Genomic Map