siRNA Details
| Gene Symbol: Olr1751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000492.1 | NP_001000492.1 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1751| Gene Symbol: Olr1751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000492.1 | NP_001000492.1 | 436 | |||