siRNA Details
| Gene Symbol: Srl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750771.3 | XP_003750819.1 | 6 | 1910 | ||
| XM_008767510.1 | XP_008765732.1 | 5 | 616 | |||
| GenBank mRNA | FQ217932.1 | - | 1 | 201 | ||
| FQ224657.1 | - | 1 | 201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srl| Gene Symbol: Srl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750771.3 | XP_003750819.1 | 6 | 1910 | ||
| XM_008767510.1 | XP_008765732.1 | 5 | 616 | |||
| GenBank mRNA | FQ217932.1 | - | 1 | 201 | ||
| FQ224657.1 | - | 1 | 201 | |||