siRNA Details
| Gene Symbol: Paqr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017377.1 | NP_001017377.1 | 1 | 378 | ||
| GenBank mRNA | BC092635.1 | - | 1 | 378 | 7315721 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Paqr4| Gene Symbol: Paqr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017377.1 | NP_001017377.1 | 1 | 378 | ||
| GenBank mRNA | BC092635.1 | - | 1 | 378 | 7315721 | |