siRNA Details
| Gene Symbol: Spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107016.1 | NP_001100486.1 | 3, 4 | 378 | ||
| GenBank mRNA | FQ233279.1 | - | 3, 4 | 250 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag7| Gene Symbol: Spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107016.1 | NP_001100486.1 | 3, 4 | 378 | ||
| GenBank mRNA | FQ233279.1 | - | 3, 4 | 250 | ||