siRNA Details
| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144991.1 | NP_001138463.1 | 8, 9 | 1457 | ||
| XM_006246562.2 | XP_006246624.2 | 8, 9 | 1454 | |||
| XM_006246563.2 | XP_006246625.2 | 8, 9 | 1454 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spns2| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144991.1 | NP_001138463.1 | 8, 9 | 1457 | ||
| XM_006246562.2 | XP_006246624.2 | 8, 9 | 1454 | |||
| XM_006246563.2 | XP_006246625.2 | 8, 9 | 1454 | |||