siRNA Details
| Gene Symbol: Cyb5d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007671.1 | NP_001007672.1 | 3 | 610 | ||
| XM_006246730.2 | XP_006246792.1 | 3 | 388 | |||
| GenBank mRNA | BC079177.1 | - | 3 | 610 | 7130324 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyb5d2| Gene Symbol: Cyb5d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007671.1 | NP_001007672.1 | 3 | 610 | ||
| XM_006246730.2 | XP_006246792.1 | 3 | 388 | |||
| GenBank mRNA | BC079177.1 | - | 3 | 610 | 7130324 | |