siRNA Details
| Gene Symbol: Tmem97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008334.1 | NP_001008335.1 | 2 | 272 | ||
| GenBank mRNA | BC085344.1 | - | 2 | 272 | 7303878 | |
| FQ214423.1 | - | 2 | 314 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Tmem97
Tmem97| Gene Symbol: Tmem97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008334.1 | NP_001008335.1 | 2 | 272 | ||
| GenBank mRNA | BC085344.1 | - | 2 | 272 | 7303878 | |
| FQ214423.1 | - | 2 | 314 | |||
 Back To Top
 Back To Top