siRNA Details
| Gene Symbol: Tmem97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008334.1 | NP_001008335.1 | 2 | 157 | ||
| GenBank mRNA | BC085344.1 | - | 2 | 157 | 7303878 | |
| FQ214423.1 | - | 2 | 199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem97| Gene Symbol: Tmem97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008334.1 | NP_001008335.1 | 2 | 157 | ||
| GenBank mRNA | BC085344.1 | - | 2 | 157 | 7303878 | |
| FQ214423.1 | - | 2 | 199 | |||