siRNA Details
| Gene Symbol: Mmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007673.1 | NP_001007674.1 | 5 | 568 | ||
| XM_006247126.1 | XP_006247188.1 | 5 | 606 | |||
| XM_006247127.2 | XP_006247189.1 | 5 | 1428 | |||
| GenBank mRNA | AF540876.1 | - | 5 | 467 | ||
| BC081965.1 | - | 5 | 568 | 7125235 | ||

Mmd
Genomic Map