siRNA Details
| Gene Symbol: Sirt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107073.1 | NP_001100543.1 | 9 | 956 | ||
| XM_006247889.2 | XP_006247951.1 | 9 | 1005 | |||
| XM_008768477.1 | XP_008766699.1 | 8 | 1077 | |||
| GenBank mRNA | BC167031.1 | - | 9 | 938 | 7099085 | |

Sirt7
Genomic Map