siRNA Details
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107107.1 | NP_001100577.2 | 7 | 1016 | ||
XM_006248177.2 | XP_006248239.1 | 7 | 1234 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107107.1 | NP_001100577.2 | 7 | 1016 | ||
XM_006248177.2 | XP_006248239.1 | 7 | 1234 |