siRNA Details
| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769048.1 | XP_008767270.1 | 15 | 1792 | ||
| GenBank mRNA | FQ211110.1 | - | 13 | 1577 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evi5l| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769048.1 | XP_008767270.1 | 15 | 1792 | ||
| GenBank mRNA | FQ211110.1 | - | 13 | 1577 | ||