siRNA Details
| Gene Symbol: Papolb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012020.2 | NP_001012020.1 | 1 | 1175 | ||
| GenBank mRNA | BC079046.1 | - | 1 | 1175 | 7115077 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Papolb| Gene Symbol: Papolb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012020.2 | NP_001012020.1 | 1 | 1175 | ||
| GenBank mRNA | BC079046.1 | - | 1 | 1175 | 7115077 | |