siRNA Details
| Gene Symbol: Mmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037217.1 | NP_001032294.1 | 3 | 344 | ||
| GenBank mRNA | BC161816.1 | - | 3 | 322 | 7320318 | |
| CB748602.1 | - | 3 | 402 | |||
| CO400908.1 | - | 3 | 321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmd2| Gene Symbol: Mmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037217.1 | NP_001032294.1 | 3 | 344 | ||
| GenBank mRNA | BC161816.1 | - | 3 | 322 | 7320318 | |
| CB748602.1 | - | 3 | 402 | |||
| CO400908.1 | - | 3 | 321 | |||