siRNA Details
| Gene Symbol: Galnt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107151.2 | NP_001100621.1 | 3 | 339 | ||
| NM_001122644.1 | NP_001116116.1 | 8 | 1651 | |||
| XM_008769333.1 | XP_008767555.1 | 8 | 1628 | |||
| XM_008769334.1 | XP_008767556.1 | 8 | 1650 | |||

Galnt9
Genomic Map