siRNA Details
Gene Symbol: Fmo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250206.2 | XP_006250268.1 | 4 | 501 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fmo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250206.2 | XP_006250268.1 | 4 | 501 |