siRNA Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107197.1 | NP_001100667.1 | 5 | 737 | ||
| XM_006250290.2 | XP_006250352.1 | 5 | 914 | |||
| XM_006250294.2 | XP_006250356.1 | 5 | 915 | |||
| XM_008769740.1 | XP_008767962.1 | 5 | 917 | |||
| XM_008769741.1 | XP_008767963.1 | 5 | 1132 | |||
| XM_008769742.1 | XP_008767964.1 | 5 | 749 | |||
| XM_008769743.1 | XP_008767965.1 | 5 | 647 | |||

Igsf9
Genomic Map