siRNA Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751237.3 | XP_003751285.1 | 2 | 1313 | ||
| XM_006249801.2 | XP_006249863.1 | 2 | 2009 | |||
| XM_223107.8 | XP_223107.5 | 2 | 977 | |||
| GenBank mRNA | FQ211467.1 | - | 2 | 646 | ||

Tmcc2
Genomic Map