siRNA Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751237.3 | XP_003751285.1 | 3 | 2148 | ||
| XM_006249801.2 | XP_006249863.1 | 3 | 2844 | |||
| XM_006249802.2 | XP_006249864.1 | 2 | 1069 | |||
| XM_006249803.2 | XP_006249865.1 | 2 | 1374 | |||
| XM_008769513.1 | XP_008767735.1 | 2 | 937 | |||
| XM_223107.8 | XP_223107.5 | 3 | 1812 | |||
| GenBank mRNA | FQ211467.1 | - | 3 | 1481 | ||

Tmcc2
Genomic Map