siRNA Details
| Gene Symbol: Rpap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013987.1 | NP_001014009.1 | 4 | 248 | ||
| XM_006250547.2 | XP_006250609.1 | 4 | 373 | |||
| XM_006250548.2 | XP_006250610.2 | 4 | 363 | |||
| XM_008769928.1 | XP_008768150.1 | 4 | 363 | |||
| XM_008769929.1 | XP_008768151.1 | 4 | 363 | |||
| XM_008769930.1 | XP_008768152.1 | 2 | 34 | |||
| GenBank mRNA | BC088426.1 | - | 4 | 248 | 7123983 | |

Rpap2
Genomic Map