siRNA Details
| Gene Symbol: Rasl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002830.2 | NP_001002830.1 | 4 | 622 | ||
| GenBank mRNA | BC083755.1 | - | 4 | 622 | 7192690 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasl11b| Gene Symbol: Rasl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002830.2 | NP_001002830.1 | 4 | 622 | ||
| GenBank mRNA | BC083755.1 | - | 4 | 622 | 7192690 | |