siRNA Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033894.1 | NP_001029066.1 | 3 | 588 | ||
| XM_006251094.1 | XP_006251156.1 | 2 | 521 | |||
| GenBank mRNA | BC104677.1 | - | 3 | 588 | 7453460 | |
| BC104718.1 | - | 2 | 528 | 7381652 | ||

Jakmip1
Genomic Map