siRNA Details
| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251532.2 | XP_006251594.1 | 9 | 2829 | ||
| XM_006251534.2 | XP_006251596.1 | 9 | 2666 | |||
| XM_008770443.1 | XP_008768665.1 | 10 | 2983 | |||
| XM_223651.8 | XP_223651.5 | 8 | 2747 | |||
| GenBank mRNA | FQ230354.1 | - | 7 | 938 | ||

Aftph
Genomic Map