siRNA Details
| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251532.2 | XP_006251594.1 | 2 | 520 | ||
| XM_006251534.2 | XP_006251596.1 | 2 | 357 | |||
| XM_008770443.1 | XP_008768665.1 | 3 | 674 | |||
| XM_223651.8 | XP_223651.5 | 2 | 519 | |||

Aftph
Genomic Map