siRNA Details
| Gene Symbol: Zfp385d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013992.1 | NP_001014014.1 | 5 | 840 | ||
| XM_008770513.1 | XP_008768735.1 | 4 | 3672 | |||
| GenBank mRNA | BC079278.1 | - | 5 | 840 | 7134172 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp385d| Gene Symbol: Zfp385d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013992.1 | NP_001014014.1 | 5 | 840 | ||
| XM_008770513.1 | XP_008768735.1 | 4 | 3672 | |||
| GenBank mRNA | BC079278.1 | - | 5 | 840 | 7134172 | |