siRNA Details
| Gene Symbol: Fezf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107251.1 | NP_001100721.1 | 2, 3 | 1136 | ||
| XM_006251737.2 | XP_006251799.1 | 4, 5 | 1399 | |||
| GenBank mRNA | BC168214.1 | - | 2, 3 | 966 | 7313369 | |

Fezf2
Genomic Map