siRNA Details
| Gene Symbol: Fezf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107251.1 | NP_001100721.1 | 2 | 788 | ||
| XM_006251737.2 | XP_006251799.1 | 4 | 1051 | |||
| GenBank mRNA | BC168214.1 | - | 2 | 618 | 7313369 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fezf2| Gene Symbol: Fezf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107251.1 | NP_001100721.1 | 2 | 788 | ||
| XM_006251737.2 | XP_006251799.1 | 4 | 1051 | |||
| GenBank mRNA | BC168214.1 | - | 2 | 618 | 7313369 | |