siRNA Details
Gene Symbol: Olr1637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000503.1 | NP_001000503.1 | 1 | 28 | ||
XM_006251869.2 | XP_006251931.1 | 2 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000503.1 | NP_001000503.1 | 1 | 28 | ||
XM_006251869.2 | XP_006251931.1 | 2 | 85 |