siRNA Details
Gene Symbol: LOC306766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014007.1 | NP_001014029.1 | 9 | 1068 | ||
GenBank mRNA | AY325163.1 | - | 715 | |||
BC085893.1 | - | 9 | 1068 | 7192798 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC306766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014007.1 | NP_001014029.1 | 9 | 1068 | ||
GenBank mRNA | AY325163.1 | - | 715 | |||
BC085893.1 | - | 9 | 1068 | 7192798 |