siRNA Details
| Gene Symbol: Nop16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047095.1 | NP_001040560.1 | 4 | 406 | ||
| GenBank mRNA | AB256043.1 | - | 4 | 406 | ||
| AM258958.1 | - | 4 | 406 | |||
| BC158562.1 | - | 4 | 410 | 9088902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nop16| Gene Symbol: Nop16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047095.1 | NP_001040560.1 | 4 | 406 | ||
| GenBank mRNA | AB256043.1 | - | 4 | 406 | ||
| AM258958.1 | - | 4 | 406 | |||
| BC158562.1 | - | 4 | 410 | 9088902 | ||